Learn More
Medchemexpress LLC USP7-IN-10 hydrochloride | 99.3% | 549.51 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY148481A1MG
Encompass_Preferred
USP7-IN-10 hydrochloride is a potent inhibitor of ubiquitin-specific protease 7 (USP7). It demonstrates an IC50 of 13.39 nM. This product is designed strictly for research applications.
  • Functions as a potent USP7 inhibitor
  • Exhibits an IC50 value of 13.39 nM
  • Intended for research use only
Catalog No. 50-003-62213