Learn More
Medchemexpress LLC USP7-IN-13 | 2305046-93-7 | 91.9% | 420.50 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY160515100MG
Encompass_Preferred
USP7-IN-13 (Compound 101) is a USP7 inhibitor with an IC50 value of 0.2-1 μM, which can be used for the study of multiple myeloma.
  • USP7 inhibitor with an IC50 value of 0.2-1 μM.
  • Useful for studying multiple myeloma.
  • Active in cancer research.
  • Targets deubiquitinase (DUBs).
  • Applicable in blood or cardio-cerebrovascular disease research.
Catalog No. 50-003-91694