Learn More
MICRO MEASUREMENT LABORATORIES INC USP729 PQ CHK STD BIMIX HIGH
Supplier:  MICRO MEASUREMENT LABORATORIES INC 061UF0125
Encompass
The MML Bi-Mix USP 729 high-concentration particle count standard #061UF-0125 provides the user with a pre-mixed Standard Preparation sample containing 5 µm and 10 µm polystyrene latex spheres (PSL) at concentrations suitable for Light Obscuration (LO) instruments equipped with an automated dilution system. MICRO MEASUREMENT LABORATORIES INC mfg# 061UF-0125
Catalog No. NC1922755