Learn More
Medchemexpress LLC USP8-IN-1 | 2477650-96-5 | 99.1% | 387.46 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY147032
Encompass_Preferred
USP8-IN-1 is a USP8 inhibitor with an IC50 of 1.9 μM. It inhibits H1975 cell growth with a GI50 of 82.04 μM. This compound is intended for research use only.
  • Inhibits USP8 with an IC50 of 1.9 μM
  • Inhibits H1975 cell growth
  • Purity of 99.07%
Catalog No. 50-003-62778