Learn More
Selleck Chemical LLC Ustekinumab-A2024-1MG-5
SDP
Supplier:  Selleck Chemical LLC A20241MG 5PC
Encompass_Preferred
Ustekinumab (CNTO 1275) is an anti-IL-12/IL-23 IgG1 human monoclonal antibody used to treat psoriasis MW 145 6 kDa
Catalog No. 50-313-4538