Learn More
Waters Corp Waters μStyragel GPC Column, 10000Å, 15–20 μm, 7.8 mm x 300 mm
Supplier: Waters Corp WAT027165
μStyragel GPC Columns are medium-efficiency columns designed for molecular weight screening. They are suitable for various analytical applications requiring precise separation.
- Chemistry: GPC
- Separation mode: SEC/GPC
- Particle substrate: Polymer
- pH range: 2 to 12
- Molecular weight range: 5000 to 600000
- Particle shape: Spherical
- Particle size: 15–20 μm
- Pore size: 10000 Å
- Format: Column
- Packing solvent: Toluene
- System: HPLC
- Technique: GPC
- Inner diameter: 7.8 mm
- Length: 300 mm
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.