Learn More
Medchemexpress LLC UT-155, Solution, 10 mM * 1 mL in DMSO
SDP
Supplier:  Medchemexpress LLC HY112895SLTN 10MM 1ML
Encompass_Preferred
UT-155 is a selective and potent androgen receptor (AR) antagonist, with a Ki of 267 nM for UT-155 binding to AR-LBD.
Catalog No. NC2942589