Learn More
Medchemexpress LLC UT-34, Solid + Solvent, 10 mM * 1 mL in DMSO, ready for reconstitution
SDP
Supplier:  Medchemexpress LLC HY136242SOLVNT10MM 1ML
Encompass_Preferred
UT-34 is a potent, selective and orally active second-generation pan-androgen receptor (AR) antagonist and degrader with IC50s of 211.7 nM, 262.4 nM and 215.7 nM for wild-type, F876L and W741L AR, respectively. UT-34 binds to ligand-binding domain (LBD) and function-1 (AF-1) domains and requires ubiquitin proteasome pathway to degrade the AR. UT-34 has anti-prostate cancer efficacy.
Catalog No. NC2942599