Learn More
Sklar Instruments Multi-Cut Utility Scissors, Sklar, Blue, 6 in., Blunt/Blunt, Angled, Serrated, Stainless Steel / Plastic. EA/1.
SDP
Supplier:  Sklar Instruments 111278
Encompass_Preferred
Multi-Cut Utility Scissors, Sklar, Blue, 6 in., Blunt/Blunt, Angled, Serrated, Stainless Steel / Plastic. Sklar Utility Scissors are a multipurpose instrument used for cutting non-sterile materials such as dressings, casts, and surgical drapes. These scissors are angled and have one serrated blade and one smooth blade. The flattened, wide, blunt tip on the serrated blade greatly reduces the likelihood of skin being nicked if the scissors are used for cutting material that is on the patient, such as clothing or bandages. They should never be used to cut tissue or sutures. One plastic finger ring is larger than the other for maximized control while cutting.
Catalog No. 50-143-7309