Learn More
Electron Microscopy Sciences Utility Shear
SDP
Supplier:  Electron Microscopy Sciences 72956
Encompass_Preferred
Multi purpose sheer with stainless steel blades and plastic handle. One blade is serrated that allows you to cut materials from wire to bone. Autoclavable to 120 degree C.
Catalog No. 50-295-79