Learn More
Medchemexpress LLC HY-117082 5mg Medchemexpress, UTL-5g CAS:646530-37-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1170825MG
Encompass_Preferred
Medchemexpress, HY-117082 5mg UTL-5g CAS:646530-37-2 UTL-5g (GBL-5g), an anti-inflammatory TNF-α inhibitor, has chemoprotective and liver radioprotective effects. UTL-5g lowers hepatotoxicity, nephrotoxicity, and myelotoxicity induced by Cisplatin through TNF-α inhibition among other factors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0898