Learn More
ESCO TECHNOLOGY INC UV-15A , UV LAMP KIT, 2&3' CAB
Supplier:  ESCO TECHNOLOGY INC 5170251
Encompass_Preferred
UV Lamp Kit, Includes One 18" UV Bulb, For 2 and 3ft cabinets, 1 per cabinet. Model UV-15A
Catalog No. NC2220624