Learn More
Andwin Scientific UV BULB SHORT WAVE F/UVGL-58
SDP
Supplier:  Andwin Scientific 36585370
Encompass
The Shortwave (254 nm) Tube, 6W is an accessory for the PL Series Compact and Handheld Ultraviolet Lamps, which are products made to perform as multipurpose tools for laboratory applications. This item can be used with UVG-47, UVG-54, UVGL-58.
Catalog No. NC9787754