Learn More
Medchemexpress LLC UV Cleavable Biotin-PEG2-Azide | 1192802-98-4 | 95.6% | C38H51N7O9S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140920100MG
Encompass_Preferred
UV Cleavable Biotin-PEG2-Azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that can participate in copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules possessing Alkyne groups. It can also engage in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Click chemistry reagent
  • Contains an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reaction (CuAAC)
  • Engages in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-56678