Learn More
Medchemexpress LLC UV Cleavable Biotin-PEG2-Azide | 1192802-98-4 | MFCD28505541 | 95.58% | C38H51N7O9S | 50 MG
Supplier: Medchemexpress LLC HY14092050MG
UV Cleavable Biotin-PEG2-Azide is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Azide group that can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
- Peg-based PROTAC linker
- Used in the synthesis of PROTACs
- Click chemistry reagent
- Azide group for Cu(I)-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups
- Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups