Learn More
Hach Company UV Lamp, Shortwave, 254 nm, 1.75"
Supplier:  Hach Company 2670800
Encompass
Shortwave (254 nm) UV lamps for field or laboratory use. Either lamp can be used in the lab with power supply while Portable Field Pack can only be used with the 1.75" lamp. Portable Field Pack requires a 9-Volt battery (not included) and has a belt clip for easy traveling. Both lamps are physically identical except for bulb and cord length (these are shorter in the 1.75" version). NOTE: Not for use in microbiological applications. UV Lamp contains mercury. Dispose according to state, local or federal regulations.
Catalog No. 50-199-6514