Learn More
Fireflysci FUV-0.2 (200-700nm) Absorbance- 0.2au (60%T)- NIST-Traceable filter for validating photometric performance of a spectrophotometer.
SDP
Supplier:  Fireflysci FUV0.2
Encompass_Preferred
The FUV-0.2 (60%T) allows calibration of photometric accuracy and stray light in the UV/VIS range (200-700nm)• Is a neutral density solid-state glass calibration standard• The following NIST wavelength points are provided for validating spectrophotometers: 250, 280, 340, 360, 400, 465, 500, 546.1, 590 and 635nm• Uncertainty of +/- 0.0022au• External dimensions of filter is 12.5x12.5x45mm• Filter is NIST traceable and produced in an ISO 17025 lab• Recommended recalibration period of 2 years
Catalog No. 50-135-4605