Learn More
Selleck Chemical LLC Uzansertib S8800-5MG
SDP
Supplier:  Selleck Chemical LLC S88005MG
Encompass_Preferred
Uzansertib (INCB053914) is a novel ATP-competitive small molecule pan-inhibitor of PIM kinases with IC50 values of 0 24 nM 30 nM and 0 12 nM for PIM1 PIM2 and PIM3 respectively in biochemical assays
Catalog No. 50-000-24596