Learn More
Medchemexpress LLC Uzansertib phosphate | 2088852-47-3 | 99.5% | 611.51 | C26H29F3N5O7P | 1 ML
SDP
Supplier:  Medchemexpress LLC HY101870B10MM 1ML
Encompass_Preferred
Uzansertib phosphate is the phosphate salt of a small-molecule kinase inhibitor, provided for research use. It is offered as a ready-to-use 10 mM solution in DMSO (1 mL) and as solid quantities, and is used in preclinical studies investigating PIM kinase-related pathways and anticancer activity.
  • Phosphate salt form for improved aqueous compatibility.
  • Supplied as 10 mM solution in DMSO (1 mL) and as solid sizes for flexibility.
  • High reported purity (~99.5%).
  • Molecular weight 611.51; chemical formula C26H29F3N5O7P.
  • Intended for laboratory research applications only.
Catalog No. 50-002-48828