Learn More
Medchemexpress LLC αvβ1 integrin-IN-1 | 1689540-62-2 | 99.5% | 558.65 g/mol | C26H34N6O6S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10044510MG
Encompass_Preferred
αvβ1 integrin-IN-1 is a small-molecule, selective inhibitor of αvβ1 integrin with potent activity (IC50 0.63 nM) and reported antifibrotic effects. It is available as the free base and as a TFA salt, supplied as solids and as a 10 mM solution in DMSO. The compound has molecular formula C26H34N6O6S, molecular weight 558.65 g/mol, and CAS 1689540-62-2, provided at high purity for research use.
  • Potent αvβ1 inhibition with IC50 0.63 nM.
  • Demonstrated antifibrotic activity in relevant assays.
  • Available as solid quantities and as a 10 MG DMSO solution (10 mM, 1 mL).
  • High purity suitable for biochemical and cell-based studies.
  • Supplied in both free base and TFA salt forms.
  • Molecular formula C26H34N6O6S; molecular weight 558.65 g/mol.
Catalog No. 50-002-14030