Learn More
TARGETMOL CHEMICALS INC V-9-M Cholecystokinin nonapept
SDP
Supplier:  TARGETMOL CHEMICALS INC TP241910MG
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. V-9-M Cholecystokinin nonapeptide (Prepro CCK Fragment V-9-M) is a precursor compound of cholecystokinin (CCK). Cholecystokinin (CCK) is a brain-intestinal peptide that stimulates gallbladder contraction and pancreatic exocrine. It can be extracted from the small intestine of dogs and cats and can cause gallbladder contraction. Purity 98.26%
Catalog No. 50-287-9400