Learn More
Meso Scale Discovery V-PLEX Ab42 Peptide 4G8 Kit
SDP
Supplier:  Meso Scale Discovery K150SLE4
Encompass
The Meso Scale Discovery V-PLEX A 42 Peptide Kit is an electrochemiluminescent immunoassay for the quantitative measurement of A 42 in human CSF and plasma It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results The kit includes a pre-coated plate detection antibody standards diluent read buffer and complete instructions With a measuring range of 50-2000 pg/mL and minimal matrix interference this kit allows reliable quantification of A 42 levels The V-PLEX platform provides a simple workflow and rapid results while using small sample volumes
Catalog No. 50-000-78326
Add to Cart
Request a Quote