Learn More
Meso Scale Discovery V-PLEX Ab42 Peptide 4G8 Kit
SDP
Supplier:  Meso Scale Discovery K150SLE1
Encompass
The Meso Scale Discovery V-PLEX A 42 Peptide Kit is an electrochemiluminescent immunoassay for the quantitative measurement of A 42 in human CSF and plasma It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results The kit includes a pre-coated plate detection antibody standards diluent read buffer and complete instructions With a measuring range of 50-2000 pg/mL and minimal matrix interference this assay delivers reliable A 42 quantification for Alzheimer s disease research The V-PLEX platform enables rapid testing of large sample sets with a streamlined and automated workflow
Catalog No. 50-000-81780