Learn More
Meso Scale Discovery V-PLEX Cytokine Panel 2 Human Kit, 1 plate
SDP
Supplier:  Meso Scale Discovery K15084D1
Encompass
The V-PLEX platform offers analytically validated singleplex and multiplex assay kits. Developed under rigorous design control, V-PLEX kits provide accurate and reproducible results with consistency from lot to lot. The V-PLEX Cytokine Panel 2 (human) Kit includes eight biomarkers that are associated with the inflammatory response and immune system regulation (Table 1). The assays can be used for quantitative measurements in the sample types listed in the “Specifications” tab below and offer detection limits (LLODs and LLOQs) in the sub- to low pg/mL range.
Catalog No. NC3072602