Learn More
Meso Scale Discovery V-PLEX GIP active Kit
SDP
Supplier:  Meso Scale Discovery K150T5D2
Encompass
The Meso Scale Discovery V-PLEX GIP Active Kit is a human GIP immunoassay kit that enables the quantitative measurement of active GIP in cell culture supernatants serum plasma and other biological samples The electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide high sensitivity wide dynamic range and rapid results The kit includes a 96-well plate coated with a GIP capture antibody detection antibody conjugated to ruthenium calibrators and reagents required to complete the assay The active GIP kit allows sensitive reproducible quantification of active GIP levels with a lower limit of detection of 0 07 pg/mL
Catalog No. 50-000-81721