Learn More
Meso Scale Discovery V-PLEX GLP-1 Total Kit
SDP
Supplier:  Meso Scale Discovery K1503PD2
Encompass
The Meso Scale Discovery V-PLEX GLP-1 Total Kit is an electrochemiluminescent immunoassay for the quantitative measurement of total glucagon-like peptide-1 (GLP-1) in human serum plasma and cell culture supernatants The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with a broad dynamic range and low sample volume requirements This kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and assay diluent The total assay time is approximately 3 5 hours
Catalog No. 50-000-82278