Learn More
Meso Scale Discovery V-PLEX Human C-Peptide Kit
SDP
Supplier:  Meso Scale Discovery K151X5D2
Encompass
The V-PLEX Human C-Peptide Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of C-peptide in human serum plasma and culture media The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a range of 80-20 000 pg/mL and takes approximately 3 5 hours to complete It uses an electrochemiluminescent detection method to provide high sensitivity and wide dynamic range The kit includes reagents for one 96-well plate which can test 40 samples in duplicate
Catalog No. 50-000-82401