Learn More
Meso Scale Discovery V-PLEX Human Cytokine 29-Plex Kit, Multiplex Immunoassay, Human, 29 analytes, Detection Limits: Sub- to low pg/mL range, Sample Types: Cell culture supernatants, Serum, Plasma, Urine

Supplier: Meso Scale Discovery K15054D2
The MESO SCALE DISCOVERY V-PLEX Human Cytokine 29-Plex Kit is a high-performance multiplex immunoassay used for the quantitative measurement of 29 human biomarkers related to inflammatory response and immune system regulation. This kit ensures accurate and reproducible results due to its robust design control, making it ideal for long-term studies across various biological sample types. This product is suitable for researchers investigating cytokines and chemokines in human samples.
- Assay type: Multiplex immunoassay
- Species reactivity: Human
- Number of analytes: 29
- Sample types: Cell culture supernatants, serum, plasma, urine
- Detection limits: Sub- to low pg/mL range
- Applications: Inflammatory response, immune system regulation
- Components: Optimized panels including Proinflammatory Panel I, Cytokine Panel I, Chemokine Panel I
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.