Learn More
Meso Scale Discovery V-PLEX Human Eotaxin-3 Kit
SDP
Supplier:  Meso Scale Discovery K151NUD2
Encompass
The Meso Scale Discovery V-PLEX Human Eotaxin-3 Kit is a sensitive immunoassay for measuring Eotaxin-3 in biological fluids It uses MSD s SULFO-TAG labels and multi-array technology requiring 50-100 L of sample The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and reagents Results are available in 3 5 hours using an MSD instrument like the MESO QuickPlex SQ 120 aiding in chemokine biology and inflammation research
Catalog No. 50-000-82268