Learn More
Meso Scale Discovery V-PLEX HUMAN IL-1 KIT
SDP
Supplier:  Meso Scale Discovery K151QPD2
Encompass
5 plates Human interleukin-1 (IL-1), also known as IL-1F2, is a 30.7 kDa pro-inflammatory cytokine that is produced by activated macrophages. IL-1β stimulates thymocyte proliferation by indu
Catalog No. NC1640887