Learn More
Meso Scale Discovery V-PLEX HUMAN IL-12/IL-23p40 KI, 96-well, Singleplex Immunoassay, Sensitivity 0.33 pg/mL, Dynamic Range 0.33 - 2,250 pg/mL, Research Use Only
SDP
Supplier:  Meso Scale Discovery K151RJD2
Encompass
The V-PLEX Human IL-12/IL-23p40 Kit is designed for the accurate and reproducible measurement of human IL-12/IL-23p40. This research-use-only kit uses the V-PLEX platform's robust design ensuring lot-to-lot consistency and the opportunity for inclusion in multiplex cytokine analyses.
  • Analytically validated performance ensures accurate and reproducible results.
  • Single-analyte measurement for focused analysis of human IL-12/IL-23p40.
  • Compatible with the V-PLEX platform for integration into broader cytokine panels.
  • Broad sample compatibility including serum, plasma, urine, and cell culture supernatant.
Catalog No. NC2032199