Learn More
Meso Scale Discovery V-PLEX Human IL-17A Kit, 5 plates, 96-well format, for measuring IL-17A, sensitivity 0.31 pg/mL, dynamic range 0.31 - 3,650 pg/mL
SDP
Supplier:  Meso Scale Discovery K151RFD2
Encompass
The MESO SCALE DISCOVERY V-PLEX Human IL-17A Kit is an analytically validated immunoassay designed for the quantitative measurement of human Interleukin-17A (IL-17A) in biological samples. This single-analyte kit provides accurate and reproducible results across different manufacturing lots and is tailored for research applications in cytokines, chemokines, and immunology/inflammation studies. The kit is adaptable for various human sample types, including serum, plasma, urine, and cell culture supernatants, ensuring broad application in research settings.
  • Measures IL-17A levels in human samples
  • Utilizes the V-PLEX platform for precision
  • 96-well plate format for ease of use
  • Compatible with multiple MSD instruments
  • Employs mouse monoclonal and goat polyclonal antibodies
  • Minimum order of 5 packs (25 plates total)
  • Dynamic range of 0.31 - 3,650 pg/mL with sensitivity of 0.31 pg/mL
  • For research use only
Catalog No. NC2879093