Learn More
Meso Scale Discovery V-PLEX Human IL-17B Kit
SDP
Supplier:  Meso Scale Discovery K151WOD2
Encompass
The V-PLEX Human IL-17B Kit from Meso Scale Discovery is a sandwich immunoassay that allows for the quantitative measurement of human interleukin 17B (IL-17B) in cell culture supernatants serum plasma and other biological fluids The assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators and reagents required to perform the assay The total assay time is approximately 3 5 hours The kit can quantify IL-17B concentrations between 1 5 pg/mL to 1000 pg/mL
Catalog No. 50-000-81893