Learn More
Meso Scale Discovery V-PLEX Human IL-17C Kit
SDP
Supplier:  Meso Scale Discovery K151WPD2
Encompass
The V-PLEX Human IL-17C Kit from Meso Scale Discovery is a sandwich immunoassay for measuring human interleukin 17C (IL-17C) in various biological fluids It uses electrochemiluminescence (ECL) detection offering high sensitivity a wide dynamic range and low sample volume requirements The kit includes a 96-well plate with capture antibody ECL-labeled detection antibody calibrators and necessary reagents The assay takes about 3 5 hours with a sensitivity of 0 3 pg/mL and an upper quantification limit of 2500 pg/mL for cell culture supernatants
Catalog No. 50-000-81897