Learn More
Meso Scale Discovery V-PLEX Human IL-9 Kit
SDP
Supplier:  Meso Scale Discovery K151XCD2
Encompass
The Meso Scale Discovery V-PLEX Human IL-9 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 9 (IL-9) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with a capture antibody specific for human IL-9 a detection antibody conjugated to an electrochemiluminescent label and calibrator diluent to generate a standard curve The assay has a sensitivity of 0 3 pg/mL and a broad dynamic range from 0 3-2500 pg/mL The total assay time is 3 5 hours This kit allows for the accurate quantitation of IL-9 levels in human samples
Catalog No. 50-000-81916