Learn More
Meso Scale Discovery V-PLEX Human IL-9 Kit 25 PL
SDP
Supplier:  Meso Scale Discovery K151XCD4
Encompass
The Meso Scale Discovery V-PLEX Human IL-9 Kit is an electrochemiluminescent immunoassay for measuring human interleukin 9 (IL-9) in various biological fluids It includes a 96-well plate with a monoclonal antibody for IL-9 a detection antibody with an electrochemiluminescent label calibrators and buffers The assay detects 3 2-10 000 pg/mL and takes about 3 5 hours It uses MSD s SECTOR Imager for high sensitivity and dynamic range with enough reagents for one 96-well plate allowing 40 samples in duplicate
Catalog No. 50-000-81796
Add to Cart
Request a Quote