Learn More
Meso Scale Discovery V-PLEX Human Leptin Kit
SDP
Supplier:  Meso Scale Discovery K151V5D1
Encompass
The V-PLEX Human Leptin Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of leptin in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with a monoclonal antibody specific for leptin a vial of SULFO-TAG labeled anti-leptin detection antibody assay diluent calibrator diluent wash buffer read buffer and a vial of lyophilized leptin calibrator The assay has a range of 3 2-10 000 pg/mL and exhibits no cross-reactivity with other cytokines The kit offers high sensitivity and requires only 25 L sample size
Catalog No. 50-000-82038