Learn More
Meso Scale Discovery V-PLEX Human MCP-1 Kit
SDP
Supplier:  Meso Scale Discovery K151NND4
Encompass
The V-PLEX Human MCP-1 Kit from Meso Scale Discovery is a multiplex immunoassay that quantitatively measures the human chemokine MCP-1 in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with capture antibodies against MCP-1 detection antibodies conjugated to an electrochemiluminescent label calibrators diluents wash buffers and other reagents required to perform the assay The kit has a detection range of 3 2-10 000 pg/mL and exhibits high sensitivity specificity and reproducibility for measuring MCP-1 levels The V-PLEX platform provides a faster and simpler workflow compared to traditional ELISA assays
Catalog No. 50-000-79152
Only null left
Add to Cart
Request a Quote