Learn More
Meso Scale Discovery V-PLEX Human MIP-1a Kit
SDP
Supplier:  Meso Scale Discovery K151NQD1
Encompass
The V-PLEX Human MIP-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the quantitative measurement of human MIP-1 in serum plasma and cell culture supernatants The kit utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluents The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-79925