Learn More
Meso Scale Discovery V-PLEX HUMAN MIP-1β KIT, 96-well, Electrochemiluminescence, MIP-1β, 0.17 - 520 pg/mL, Serum, Plasma, Urine, Cell Culture Supernatant

Supplier: Meso Scale Discovery K151NRD4
The MESO SCALE DISCOVERY V-PLEX Human MIP-1β Kit is a validated singleplex assay for accurately measuring human Macrophage Inflammatory Protein 1-beta. Built on the V-PLEX platform, this kit guarantees consistent lot-to-lot performance and is designed for use in a variety of biological samples, making it an essential tool for scientific research.
- Analyte: MIP-1β
- Species: Human
- Platform: V-PLEX
- Detection method: Electrochemiluminescence
- Sensitivity: 0.17 pg/mL
- Dynamic range: 0.17 - 520 pg/mL
- Sample types: Serum, Plasma, Urine, Cell Culture Supernatant
- Capture antibody: Mouse Monoclonal
- Detection antibody: Mouse Monoclonal
- Associated gene ID: 6351
- Associated UniProt ID: P13236
- Primary application areas: Cytokines & Chemokines, Immunology/Inflammation
- Storage condition: 2-8 °C
- For research use only. Not for diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.