Learn More
Meso Scale Discovery V-PLEX Human PP Kit
SDP
Supplier:  Meso Scale Discovery K151Y5D1
Encompass
The V-PLEX Human PP Kit from Meso Scale Discovery is a multiplex immunoassay for measuring Pancreatic Polypeptide (PP) in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The assay requires 50 L sample volume and takes 3 5 hours The kit includes a 96-well plate ECL-labeled detection antibody calibrators buffers plate seals and instructions Validated for research use only not for diagnostics
Catalog No. 50-000-79819