Learn More
Meso Scale Discovery V-PLEX Human SAA Kit
SDP
Supplier:  Meso Scale Discovery K151SSD4
Encompass
The V-PLEX Human SAA Kit from Meso Scale Discovery is an immunoassay that quantitatively measures serum amyloid A (SAA) in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and rapid results The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators and reagents required to perform the assay SAA levels can be accurately quantified in under 3 hours This assay kit allows for the measurement of SAA an important biomarker of inflammation
Catalog No. 50-000-80542
Only null left
Add to Cart
Request a Quote