Learn More
Meso Scale Discovery V-PLEX Human VCAM-1 Kit
SDP
Supplier:  Meso Scale Discovery K151SRD1
Encompass
The V-PLEX Human VCAM-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the quantitative measurement of human VCAM-1 in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies against human VCAM-1 detection antibodies conjugated to an electrochemiluminescent label calibrator diluent wash buffer read buffer and a vial of lyophilized calibrator that must be reconstituted before use The assay has a detection range of 2 4-10 000 pg/mL and exhibits minimal cross-reactivity to related analytes The kit components are for research use only and not intended for diagnostic procedures
Catalog No. 50-000-78574