Learn More
Meso Scale Discovery V-PLEX Human VEGF-C Kit
SDP
Supplier:  Meso Scale Discovery K151LTD1
Encompass
The V-PLEX Human VEGF-C Kit from Meso Scale Discovery is a sandwich immunoassay that quantitatively measures human VEGF-C in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with a VEGF-C capture antibody a VEGF-C detection antibody calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 31 pg/mL and an assay range from 0 31-2000 pg/mL The total assay time is 3 5 hours This kit allows for the accurate and sensitive measurement of human VEGF-C levels
Catalog No. 50-000-79962