Learn More
Meso Scale Discovery V-PLEX Mouse IL-15 Kit
SDP
Supplier:  Meso Scale Discovery K152RDD2
Encompass
The Meso Scale Discovery V-PLEX Mouse IL-15 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 15 (IL-15) in cell culture supernatants serum plasma and tissue homogenates The assay utilizes MSD s proprietary SULFO-TAG labels which emit light when electrochemically stimulated via carbon electrodes in the bottom of each microwell This kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium calibrators and diluent The assay has a sensitivity of 1 3 pg/mL and a broad dynamic range from 1 3-10 000 pg/mL
Catalog No. 50-000-82148