Learn More
Meso Scale Discovery V-PLEX Mouse IL-5 Kit
SDP
Supplier:  Meso Scale Discovery K152QSD2
Encompass
The Meso Scale Discovery V-PLEX Mouse IL-5 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 5 (IL-5) in cell culture supernatants serum and plasma The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and assay diluent The assay has a sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL It can detect natural and recombinant mouse IL-5 with a sample volume of 50 L The total assay time is 3 5 hours This kit allows for the accurate and sensitive quantification of mouse IL-5 levels
Catalog No. 50-000-78435