Learn More
Meso Scale Discovery V-PLEX Mouse IL-5 Kit
SDP
Supplier:  Meso Scale Discovery K152QSD4
Encompass
The Meso Scale Discovery V-PLEX Mouse IL-5 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 5 (IL-5) in cell culture supernatants serum plasma and tissue lysates The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 1 53-2500 pg/mL and takes approximately 3 5 hours to complete This highly sensitive and reproducible kit allows for the accurate quantification of mouse IL-5
Catalog No. 50-000-79103
Add to Cart
Request a Quote