Learn More
Meso Scale Discovery V-PLEX Mouse IL-6 Kit
SDP
Supplier:  Meso Scale Discovery K152QXD4
Encompass
The Meso Scale Discovery V-PLEX Mouse IL-6 Kit is an electrochemiluminescent immunoassay for quantifying mouse interleukin 6 (IL-6) in various samples It includes a 96-well plate with capture antibody detection antibody with an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay detects 0 3-2500 pg/mL takes about 3 5 hours and uses the MSD QuickPlex SQ 120 for reading It is highly sensitive requires only 50 L of sample and does not cross-react with other cytokines
Catalog No. 50-000-81332
Add to Cart
Request a Quote