Learn More
Meso Scale Discovery V-PLEX Mouse MCP-1 Kit
SDP
Supplier:  Meso Scale Discovery K152NND4
Encompass
The V-PLEX Mouse MCP-1 Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of monocyte chemoattractant protein 1 (MCP-1) in mouse biological samples such as serum plasma and cell culture supernatants The assay utilizes MSD s proprietary V-PLEX technology is highly sensitive has a broad dynamic range of 24-20 000 pg/mL and shows minimal cross-reactivity with other cytokines The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer diluent and calibrator The total assay time is 3 5 hours
Catalog No. 50-000-81408
Only null left
Add to Cart
Request a Quote