Learn More
Meso Scale Discovery V-PLEX Mouse MIP-2 Kit
SDP
Supplier:  Meso Scale Discovery K152QCD4
Encompass
The V-PLEX Mouse MIP-2 Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of mouse MIP-2 in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and plate seals The electrochemiluminescent assay has a sensitivity of 1 3 pg/mL and an upper limit of quantification of 10 000 pg/mL The total assay time is 3 5 hours This quantitative immunoassay allows for the accurate measurement of mouse MIP-2 levels
Catalog No. 50-000-82226
Add to Cart
Request a Quote